missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endosulfan in Methanol 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
10 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 115-29-7 |
| Molecular Formula | C9H6Cl6O3S |
| Concentration | 10 μg/mL |
| InChI Key | RDYMFSUJUZBWLH-UHFFFAOYSA-N |
| SMILES | C1C2C(COS(=O)O1)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 406.904 |
| PubChem CID | 3224 |
| Quantity | 10 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction