missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine in Acetonitirle 10μg/mL, Fisher Chemical™

Product Code. 12946583 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12946583 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12946583 Supplier Fisher Chemical Supplier No. P822500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 122-39-4
Molecular Formula C12H11N
Concentration 10 μg/mL
UN Number 1648
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2
IUPAC Name N-phenylaniline
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 169.227
PubChem CID 11487
Quantity 10 mL
ChEBI CHEBI:4640
Chemical Name or Material Diphenylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Diphenylamine 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.