missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Decafluorotriphenylphosphine in DCM 100μg/mL, Fisher Chemical™

Product Code. 12992654 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12992654 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12992654 Supplier Fisher Chemical Supplier No. P816180

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 5074-71-5
Molecular Formula C18H5F10P
Concentration 100 μg/mL
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)(phenyl)phosphane
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 442.20
PubChem CID 78757
Quantity 1 mL
Chemical Name or Material Decafluorotriphenylphosphine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.