missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrysene in Acetone 10μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 218-01-9 |
| Molecular Formula | C18H12 |
| Concentration | 10 μg/mL |
| UN Number | 1090 |
| InChI Key | WDECIBYCCFPHNR-UHFFFAOYSA-N |
| SMILES | C1=CC=C2C(=C1)C=CC3=C2C=CC4=CC=CC=C43 |
| IUPAC Name | chrysene |
| Solvent or Matrix | Acetone |
| Molecular Weight (g/mol) | 228.294 |
| Show More |
Safety and Handling
Product Identifier
- Standard solution Chrysene 10ug/ml in acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction