missing translation for 'onlineSavingsMsg'
Learn More

Chlortoluron in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12932534 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12932534 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12932534 Supplier Fisher Chemical Supplier No. P812930
  • ONLINE EXCLUSIVE PRICE
    £35.90
    £35.25 / 1mL
    Save £0.65
    1% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Especificaciones

Packaging Ampoule(s)
CAS 15545-48-9
Molecular Formula C10H13ClN2O
Concentration 100 μg/mL
MDL Number MFCD00018275
UN Number 1648
InChI Key JXCGFZXSOMJFOA-UHFFFAOYSA-N
SMILES CN(C)C(=O)NC1=CC=C(C)C(Cl)=C1
IUPAC Name 3-(3-chloro-4-methylphenyl)-1,1-dimethylurea
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 212.68
PubChem CID 27375
Quantity 1 mL
ChEBI CHEBI:81981
Chemical Name or Material Chlortoluron
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Standard solution Chlortoluron 100ug/ml in acetonitrile
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.