missing translation for 'onlineSavingsMsg'
Learn More

Chlorpyrifos in Acetonitrile 100μg/mL, Fisher Chemical™

Product Code. 12912524 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12912524 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12912524 Supplier Fisher Chemical Supplier No. P812630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 2921-88-2
Molecular Formula C9H11Cl3NO3PS
Concentration 100 μg/mL
UN Number 1648
InChI Key SBPBAQFWLVIOKP-UHFFFAOYSA-N
SMILES CCOP(=S)(OCC)OC1=NC(=C(C=C1Cl)Cl)Cl
IUPAC Name diethoxy-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-$l^{5}-phosphane
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 350.575
PubChem CID 2730
Quantity 1 mL
ChEBI CHEBI:34631
Chemical Name or Material Chlorpyrifos
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Chlorpyrifos 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H410-Very toxic to aquatic life with long lasting effects.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.