missing translation for 'onlineSavingsMsg'
Learn More

Chlormequat Chloride in Acetonitrile 10μg/mL, Fisher Chemical™

Product Code. 12992414 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 12992414

Brand: Fisher Chemical P810660

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 999-81-5
Molecular Formula C5H13Cl2N
Concentration 10 μg/mL
UN Number 1648
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
SMILES C[N+](C)(C)CCCl.[Cl-]
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 158.066
PubChem CID 13836
Quantity 10 mL
Chemical Name or Material Chlormequat Chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Chlormequat chloride,2-chloroethyltrimethylammonium chloride 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.