missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Carbofuran in Cyclohexane 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 1563-66-2 |
| Molecular Formula | C12H15NO3 |
| Concentration | 100 μg/mL |
| UN Number | 1145 |
| InChI Key | DUEPRVBVGDRKAG-UHFFFAOYSA-N |
| SMILES | CC1(CC2=C(O1)C(=CC=C2)OC(=O)NC)C |
| IUPAC Name | (2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate |
| Solvent or Matrix | Cyclohexane |
| Molecular Weight (g/mol) | 221.256 |
| Show More |
Safety and Handling
Product Identifier
- Standard solution Carbofuran 100ug/ml in cyclohexane
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction