missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Carbofuran in Acetonitrile 100μg/mL, Fisher Chemical™

Product Code. 12979763 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12979763

Brand: Fisher Chemical P809800

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 1563-66-2
Molecular Formula C12H15NO3
Concentration 100 μg/mL
UN Number 1648
InChI Key DUEPRVBVGDRKAG-UHFFFAOYSA-N
SMILES CC1(CC2=C(O1)C(=CC=C2)OC(=O)NC)C
IUPAC Name (2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 221.256
PubChem CID 2566
Quantity 1 mL
ChEBI CHEBI:34611
Chemical Name or Material Carbofuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Carbofuran,2,3-dihydro-2,2-dimethylbenzofuran-7-yl N-methylcarbamate 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.