missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Carbofuran in Acetonitrile 10μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 1563-66-2 |
| Molecular Formula | C12H15NO3 |
| Concentration | 10 μg/mL |
| UN Number | 1648 |
| InChI Key | DUEPRVBVGDRKAG-UHFFFAOYSA-N |
| SMILES | CC1(CC2=C(O1)C(=CC=C2)OC(=O)NC)C |
| IUPAC Name | (2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate |
| Solvent or Matrix | Acetonitrile |
| Molecular Weight (g/mol) | 221.256 |
| Show More |
Safety and Handling
Product Identifier
- Standard solution Carbofuran,2,3-dihydro-2,2-dimethylbenzofuran-7-yl N-methylcarbamate 10ug/ml in acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H311-Toxic in contact with skin.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction