missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Carbazole in DCM 100μg/mL, Fisher Chemical™

Product Code. 12919763 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12919763

Brand: Fisher Chemical P809570

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 86-74-8
Molecular Formula C12H9N
Concentration 100 μg/mL
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2
IUPAC Name 9H-carbazole
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 167.21
PubChem CID 6854
Quantity 1 mL
ChEBI CHEBI:27543
Chemical Name or Material Carbazole
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.