missing translation for 'onlineSavingsMsg'
Learn More

Captan in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA

Product Code. 12929753 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12929753

Brand: Fisher Chemical P809330

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 1527-98-6
Molecular Formula C10H12N4O4
Concentration 100 μg/mL
MDL Number MFCD00191327
UN Number 1648
InChI Key IKGRHEWIFBFXPP-IZZDOVSWSA-N
SMILES CCC\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
IUPAC Name (1E)-1-butylidene-2-(2,4-dinitrophenyl)hydrazine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 252.23
PubChem CID 9601272
Quantity 1 mL
Chemical Name or Material Captan
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Captan,1,2,3,6-tetrahydro-N-(trichloromethylthio)phthalimide 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.