missing translation for 'onlineSavingsMsg'
Learn More

Beta-BHC in Toluene 100μg/mL, Fisher Chemical™

Product Code. 12946833 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12946833 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12946833 Supplier Fisher Chemical Supplier No. P827590

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 319-85-7
Molecular Formula C6H6Cl6
Concentration 100 μg/mL
InChI Key JLYXXMFPNIAWKQ-UHFFFAOYSA-N
SMILES C1(C(C(C(C(C1Cl)Cl)Cl)Cl)Cl)Cl
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
Solvent or Matrix Toluene
Molecular Weight (g/mol) 290.814
PubChem CID 727
Quantity 1 mL
ChEBI CHEBI:24536
Chemical Name or Material Beta-HCH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.