missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Benzo(g,h,i)perylene in DCM 10μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 191-24-2 |
| Molecular Formula | C22H12 |
| Concentration | 10 μg/mL |
| InChI Key | GYFAGKUZYNFMBN-UHFFFAOYSA-N |
| SMILES | C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 |
| Solvent or Matrix | Dichloromethane |
| Molecular Weight (g/mol) | 276.338 |
| PubChem CID | 9117 |
| Quantity | 10 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction