missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1260 in Iso-octane 10μg/mL, Fisher Chemical™

Product Code. 12999533 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Packungsgröße:
10mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 12999533

Marke: Fisher Chemical P804840

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Spezifikation

Packaging Ampoule
CAS 11096-82-5
Physical Form Liquid
Molecular Formula C12H4Cl6
Concentration 10 μg/mL
UN Number 1262
InChI Key BTAGRXWGMYTPBY-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC(=C(C(=C1C2=C(C(=C(C=C2)Cl)Cl)Cl)Cl)Cl)Cl
IUPAC Name 1,2,3-trichloro-4-(2,3,4-trichlorophenyl)benzene
Solvent or Matrix Iso-octane
Molecular Weight (g/mol) 360.864
PubChem CID 38018
Quantity 10 mL
Chemical Name or Material Aroclor 1260
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • ORGANIC SINGLE COMPONENT STANDARD SOLUTION 10ug/ml Aroclor 1260 in Iso-octane (P804840)
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt