missing translation for 'onlineSavingsMsg'
Learn More

Anthracene in DCM 100μg/mL, Fisher Chemical™

Product Code. 12929523 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12929523 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12929523 Supplier Fisher Chemical Supplier No. P804310

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 120-12-7
Physical Form Liquid
Molecular Formula C14H10
Concentration 100 μg/mL
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1
IUPAC Name anthracene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 178.23
PubChem CID 8418
Quantity 1 mL
ChEBI CHEBI:35298
Chemical Name or Material Anthracene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.