missing translation for 'onlineSavingsMsg'
Learn More

Metabolite Alpha-Endosulfan in Methanol 10μg/mL, Fisher Chemical™

Product Code. 12946613 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12946613 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12946613 Supplier Fisher Chemical Supplier No. P823120

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 959-98-8
Molecular Formula C9H6Cl6O3S
Concentration 10 μg/mL
InChI Key RDYMFSUJUZBWLH-AZVNHNRSSA-N
SMILES C1C2C(COS(=O)O1)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl
Solvent or Matrix Methanol
Molecular Weight (g/mol) 406.904
PubChem CID 12309460
Quantity 10 mL
Chemical Name or Material Endosulfan-alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.