missing translation for 'onlineSavingsMsg'
Learn More

Alpha,Alpha,Alpha-Trifluorotoluene in Methanol 200μg/mL, Fisher Chemical™

Product Code. 12976003 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12976003 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12976003 Supplier Fisher Chemical Supplier No. F113071

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 98-08-8
Molecular Formula C7H5F3
Concentration 200 μg/mL
UN Number 1230
InChI Key GETTZEONDQJALK-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC=C(C=C1)C(F)(F)F
IUPAC Name trifluoromethylbenzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 146.112
PubChem CID 7368
Quantity 1 mL
ChEBI CHEBI:36810
Chemical Name or Material Alpha,Alpha,Alpha-Trifluorotoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.