missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12973134 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12973134 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12973134 Supplier Fisher Chemical Supplier No. P873880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 98-86-2
Physical Form Liquid
Molecular Formula C8H8O
Concentration 100 μg/mL
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC(=O)C1=CC=CC=C1
IUPAC Name 1-phenylethanone
Solvent or Matrix Methanol
Molecular Weight (g/mol) 120.151
PubChem CID 7410
Quantity 5 mL
ChEBI CHEBI:27632
Chemical Name or Material Acetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.