missing translation for 'onlineSavingsMsg'
Learn More

Acetone-DNPH in Acetonitrile 100μg/mL, Fisher Chemical™

Product Code. 12996263 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12996263 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996263 Supplier Fisher Chemical Supplier No. P802930

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Spécification

Packaging Ampoule
CAS 1567-89-1
Physical Form Liquid
Molecular Formula C9H10N4O4
Concentration 100 μg/mL
MDL Number MFCD00137172
UN Number 1648
InChI Key YGIXYAIGWMAGIB-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC(C)=NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
IUPAC Name 1-(2,4-dinitrophenyl)-2-(propan-2-ylidene)hydrazine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 238.20
PubChem CID 74069
Quantity 1 mL
Chemical Name or Material Acetone-DNPH
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Standard solution Acetone-DNPH 100ug/ml in acetonitrile
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.