missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthylene in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12996243 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12996243 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12996243 Supplier Fisher Chemical Supplier No. P802550

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 208-96-8
Physical Form Liquid
Molecular Formula C12H8
Concentration 100 μg/mL
MDL Number MFCD00003806
InChI Key HXGDTGSAIMULJN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC2=C3C1=CC=CC3=CC=C2
IUPAC Name acenaphthylene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 152.20
PubChem CID 9161
Quantity 1 mL
ChEBI CHEBI:33081
Chemical Name or Material Acenaphthylene
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.