missing translation for 'onlineSavingsMsg'
Learn More

Metabolite 4,4'-Dibromobiphenyl in DCM 100μg/mL, Fisher Chemical™

Product Code. 12955304 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12955304 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12955304 Supplier Fisher Chemical Supplier No. P817190

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule(s)
CAS 92-86-4
Molecular Formula C12H8Br2
Concentration 100 μg/mL
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
IUPAC Name 4,4'-dibromo-1,1'-biphenyl
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 312.00
PubChem CID 7110
Quantity 1 mL
Chemical Name or Material 4,4'-Dibromobiphenyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.