missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-DDD (TDE) in Methanol 10μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 10μg/mL |
CAS | 72-54-8 |
Packaging | Ampoule(s) |
Quantity | 10 mL |
Molecular Formula | C14H10Cl4 |
UN Number | 1230 |
InChI Key | AHJKRLASYNVKDZ-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)Cl)Cl |
IUPAC Name | 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene |
Molecular Weight (g/mol) | 320.034 |
Show More |
Safety and Handling
Product Identifier
- Standard solution 4,4'-DDD (TDE) 10ug/ml in methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'