missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,1,3,3-Tetramethylbutyl)phenol in Toluene 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 140-66-9 |
| Molecular Formula | C14H22O |
| Concentration | 100 μg/mL |
| InChI Key | ISAVYTVYFVQUDY-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | CC(C)(C)CC(C)(C)C1=CC=C(C=C1)O |
| IUPAC Name | 4-(2,4,4-trimethylpentan-2-yl)phenol |
| Solvent or Matrix | Toluene |
| Molecular Weight (g/mol) | 206.329 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction