missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic acid in Water 1104μg/mL, Fisher Chemical™

Product Code. 12986143 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12986143 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12986143 Supplier Fisher Chemical Supplier No. F128991

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 118-91-2
Molecular Formula C7H5ClO2
Concentration 1104 μg/mL
InChI Key IKCLCGXPQILATA-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC=C(C(=C1)C(=O)O)Cl
IUPAC Name 2-chlorobenzoic acid
Solvent or Matrix Water
Molecular Weight (g/mol) 156.565
PubChem CID 8374
Quantity 1 mL
ChEBI CHEBI:30793
Chemical Name or Material 2-Chlorobenzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.