missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol in 2-propanol 1000μg/mL, Fisher Chemical™

Product Code. 12986093 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12986093

Brand: Fisher Chemical F116531

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 615-58-7
Molecular Formula C6H4Br2O
Concentration 1000 μg/mL
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=CC(=C(C=C1Br)Br)O
IUPAC Name 2,4-dibromophenol
Solvent or Matrix 2- Propanol
Molecular Weight (g/mol) 251.905
PubChem CID 12005
Quantity 1 mL
ChEBI CHEBI:34238
Chemical Name or Material 2,4-Dibromophenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.