missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4'-DDT in Methanol 10μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 10μg/mL |
CAS | 789-02-6 |
Packaging | Ampoule(s) |
Quantity | 10 mL |
Molecular Formula | C14H9Cl5 |
InChI Key | CVUGPAFCQJIYDT-UHFFFAOYSA-N |
SMILES | C1=CC=C(C(=C1)C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl |
IUPAC Name | 1-chloro-2-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene |
Molecular Weight (g/mol) | 354.476 |
PubChem CID | 13089 |
Show More |
Safety and Handling
Product Identifier
- 2,4 prime-DDT [CAS:789-02-6] 10 ug/ml in Methanol (P815870)
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'