missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrachloro-m-xylene in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12980154 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12980154

Brand: Fisher Chemical P853470

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 877-09-8
Physical Form Liquid
Molecular Formula C8H6Cl4
Concentration 100 μg/mL
InChI Key NTUBJKOTTSFEEV-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC1=C(C(=C(C(=C1Cl)Cl)Cl)C)Cl
IUPAC Name 1,2,3,5-tetrachloro-4,6-dimethylbenzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 243.936
PubChem CID 70139
Quantity 1 mL
Chemical Name or Material 2,4,5,6-Tetrachloro-m-xylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato