missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5,6-Tetrachloro-m-xylene in Acetone 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 877-09-8 |
| Physical Form | Liquid |
| Molecular Formula | C8H6Cl4 |
| Concentration | 100 μg/mL |
| UN Number | 1090 |
| InChI Key | NTUBJKOTTSFEEV-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | CC1=C(C(=C(C(=C1Cl)Cl)Cl)C)Cl |
| IUPAC Name | 1,2,3,5-tetrachloro-4,6-dimethylbenzene |
| Show More |
Safety and Handling
Product Identifier
- Standard solution 2,4,5,6-Tetrachloro-m-xylene 100ug/ml in acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction