missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12995354 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12995354 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12995354 Supplier Fisher Chemical Supplier No. P818160

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule(s)
CAS 541-73-1
Molecular Formula C6H4Cl2
Concentration 100 μg/mL
UN Number 1230
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)Cl
IUPAC Name 1,3-dichlorobenzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 146.998
PubChem CID 10943
Quantity 1 mL
ChEBI CHEBI:36693
Chemical Name or Material 1,3-Dichlorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution 1,3-Dichlorobenzene 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.