missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trichlorobenzene in Methanol 10μg/mL, Fisher Chemical™

Product Code. 12920284 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12920284 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12920284 Supplier Fisher Chemical Supplier No. P855730

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 108-70-3
Physical Form Liquid
Molecular Formula C6H3Cl3
Concentration 10 μg/mL
MDL Number MFCD00000585
UN Number 1230
InChI Key XKEFYDZQGKAQCN-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES ClC1=CC(Cl)=CC(Cl)=C1
IUPAC Name 1,3,5-trichlorobenzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 181.44
PubChem CID 7950
Quantity 10 mL
ChEBI CHEBI:49916
Chemical Name or Material 1,3,5-Trichlorobenzene
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Standard solution 1,3,5-Trichlorobenzene 10ug/ml in methanol
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.