missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrachlorobenzene in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12960144 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
12960144 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 12960144

Brand: Fisher Chemical P853270

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Ampoule
CAS 95-94-3
Physical Form Liquid
Molecular Formula C6H2Cl4
Concentration 100 μg/mL
UN Number 1230
InChI Key JHBKHLUZVFWLAG-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)Cl
IUPAC Name 1,2,4,5-tetrachlorobenzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 215.882
PubChem CID 7270
Quantity 1 mL
ChEBI CHEBI:36697
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution 1,2,4,5-Tetrachlorobenzene 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.