missing translation for 'onlineSavingsMsg'
Learn More

Meta-Phosphoric Acid, ACS Reagent, Honeywell Fluka™

Product Code. 15602440 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
6 x 500 g
Unit Size:
100g
2.5kg
500g
5g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15602440 100 g 100g
15632440 5 g 5g
15622440 500 g 500g
15612440 2.5 kg 2.5kg
15642440 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15602440 Supplier Honeywell-Fluka Supplier No. 239275100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Quantity 100 g
Linear Formula (HPO3)n
UN Number 3260
Grade ACS Reagent
Chemical Name or Material Meta-Phosphoric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.