missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MET, Rabbit anti-Human, PE, Clone: 6F11, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB18955.100Reactions
This item is not returnable.
View return policy
Specifications
| MET | |
| Monoclonal | |
| PE | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| AUTS9/HGFR/RCCP2/c-Met | |
| Rabbit | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 6F11 | |
| Flow Cytometry (5 uL/106 cells or 0.05 ug/mL) The optimal working dilution should be determined by the end user. | |
| met proto-oncogene (hepatocyte growth factor receptor) | |
| MET | |
| A synthetic phosphopeptide corresponding to residues surrounding Y1234/Y1235 of human MET. | |
| RUO | |
| 4233 | |
| Store at 2-8°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction