Learn More
Beckman Coulter Life Sciences MET ONE HHPC 2+ Handheld
Product Code. 31777222 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Particle Size:
0.5 to 5 μm
Unit Size:
Each
Product Code. Particle Size Unit Size
31777222 0.5 to 5 μm Each
1 options
Product Code. 31777222 Supplier Beckman Coulter Life Sciences Supplier No. 208931003
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed as an affordable tool for the routine monitoring of controlled environments to Cleanroom Classifications.
  • Designed for regulated and semi regulated production environments adhering to GMP principles
  • Suitable for aseptically packaged food and beverage filling lines
  • Used in hospital labs and formulating pharmacies
  • MET ONE HHPC 2+ is frequently chosen for these applications
Specifications
Product Type Particle Counter
Display Type TFT Color LCD
Particle Size 0.5 to 5 μm
Designed for spot checks and troubleshooting; not intended to replace continuous cleanroom monitoring systems.