missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beckman Coulter Life Sciences MET ONE GMP Cleanroom Routine Environmental Air Particle Counter, 28.3 L/min.
Description
- Portable air particle counter supports GMP cleanroom environmental monitoring and improves data integrity.
- Lighter and quieter design compared to previous models.
- Allows import of SOP routine monitoring maps and sampling configurations directly into the counter.
- SOP configuration sampling maps and version control managed via web browser.
- Administrator manages SOP versions using electronic signatures.
- Sampling map guides users through daily monitoring program.
- Completed samples are indicated on screen for easy tracking.
- Results can be reviewed and approved remotely by a supervisor via web browser.
- Approved reports include electronic signature and can be exported in secure electronic format.
Specifications
Specifications
| Product Type | Particle Counter |
| Display Type | TFT Color LCD |
| Grid Type | Gridless |
| Particle Size | 0.3 μm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?