missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Met-Enkephalin

Product Code. 15433789
Change view
Click to view available options
Quantity:
25 mg
50 mg
125 mg
Unit Size:
25mg
50mg
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433789 50 mg 50mg
15423789 25 mg 25mg
15413789 125 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433789 Supplier Thermo Scientific Chemicals Supplier No. J61154.MF
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 58569-55-4573.665 g/mol

Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58569-55-4

Specifications

Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 573.665
Product Type Met-Enkephalin
Quantity 50 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.