missing translation for 'onlineSavingsMsg'
Learn More

Mesterolone, TRC

Product Code. 30005608
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005608 50 mg 50mg
30005830 100 mg 100mg
30009872 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
Product Code. 30005608 Supplier Toronto Research Chemicals Supplier No. TRCM22580050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1424-00-6
Molecular Formula C20 H32 O2
Molecular Weight (g/mol) 304.47
Synonym Androstan-3-one, 17-hydroxy-1-methyl-, (1α,5α,17β)-, 5α-Androstan-3-one, 17β-hydroxy-1α-methyl- (6CI,7CI,8CI), (1α,5α,17β)-17-Hydroxy-1-methylandrostan-3-one, 17β-Hydroxy-1α-methyl-5α-androstan-3-one, 1α-Methyl-17β-hydroxy-5α-androstan-3-one, 1α-Methyl-5α-androstan-17β-ol-3-one, 1α-Methyl-5α-dihydrotestosterone, Androviron, MEST, Mesteranum, Mesterolone, Mestoranum, NSC 75054, Proviron, Provirone 25, SH 723, Testiwop
IUPAC Name (1S,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-1,10,13-trimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@H]1CC(=O)C[C@@H]2CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@@H]3[C@@]12C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H32O2/c1-12-10-14(21)11-13-4-5-15-16-6-7-18(22)19(16,2)9-8-17(15)20(12,13)3/h12-13,15-18,22H,4-11H2,1-3H3/t12-,13-,15-,16-,17-,18-,19-,20-/m0/s1
Formula Weight 304.2402
Chemical Name or Material Mesterolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.