missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEST Antibody (5E0), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42997
This item is not returnable.
View return policy
Description
MEST Monoclonal antibody specifically detects MEST in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| MEST | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 5 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| DKFZp686L18234, EC 3.-, mesoderm specific transcript (mouse) homolog, mesoderm specific transcript homolog (mouse), mesoderm-specific transcript homolog protein, MGC111102, MGC8703, paternally expressed gene 1, PEG1Paternally-expressed gene 1 protein | |
| mesoderm specific transcript homolog(Mouse) - (Uniprot# Q5EB52) | |
| 100 μg | |
| Cell Biology | |
| 4232 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction