missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MESP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575751
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).
Specifications
| MESP2 | |
| Polyclonal | |
| Unconjugated | |
| Mesp2 | |
| bHLHc6; Class C basic helix-loop-helix protein 6; mesoderm posterior 2; mesoderm posterior 2 homolog; mesoderm posterior basic helix-loop-helix transcription factor 2; mesoderm posterior bHLH transcription factor 2; mesoderm posterior protein 2; Mesp2; SCDO2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 145873, 17293, 293046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O08574, Q0VG99 | |
| Mesp2 | |
| Synthetic peptide corresponding to residues in Human MESP2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction