missing translation for 'onlineSavingsMsg'
Learn More

Mesotrione, TRC

Product Code. 30466999
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466999 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466999 Supplier Toronto Research Chemicals Supplier No. TRCM22576510G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesotrione
CAS 104206-82-8
Molecular Formula C14 H13 N O7 S
Quantity 10 g
Synonym 2-[4-(Methylsulfonyl)-2-nitrobenzoyl]-1,3-cyclohexanedione, Callisto, Mesotrione, Tenacity, Tenacity (herbicide), Xiaohuangcaotong, ZA 1296
InChI Formula InChI=1S/C14H13NO7S/c1-23(21,22)8-5-6-9(10(7-8)15(19)20)14(18)13-11(16)3-2-4-12(13)17/h5-7,13H,2-4H2,1H3
SMILES CS(=O)(=O)c1ccc(C(=O)C2C(=O)CCCC2=O)c(c1)[N+](=O)[O-]
IUPAC Name 2-(4-methylsulfonyl-2-nitrobenzoyl)cyclohexane-1,3-dione
Molecular Weight (g/mol) 339.32
Formula Weight 339.0413
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.