missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin, Mouse anti-Human,Mouse,Rat, Clone: MSLN/2131, Novus Biologicals™
Mouse Monoclonal Antibody
£224.00 - £403.00
Specifications
| Antigen | Mesothelin |
|---|---|
| Clone | MSLN/2131 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18213138
|
Novus Biologicals
NBP2-79724-20UG |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18273968
|
Novus Biologicals
NBP2-79724-100UG |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Mesothelin | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| MSLN | |
| IgG2b κ | |
| Protein A or G purified |
| MSLN/2131 | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q13421, Q13421 | |
| 10232 | |
| Recombinant fragment (around aa 273-407) of human Mesothelin protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title