missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mesothelin Monoclonal Antibody (C6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543664

 View more versions of this product

Product Code. 17684116

  • £178.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Originally identified as a differentiation antigen of mesotheliomas, ovarian cystadenocarcinomas, and pancreatic adenocarcinomas, Mesothelin is a glycosylphosphatidylinositol (GPI)-anchored, cell-surface glycoprotein predominantly secreted by cells of the mesothelium. Although Mesothelin is expressed at restricted levels by normal mesothelial cells of the pleural,pericardial, and peritoneal membranes, aberrant expression has been documented in the aforementioned cancers, as well as in endometriod uterine adenocarcinomas and squamous cell carcinomas of the esophagus, stomach, lung, and cervix. Proteolytic cleavage of Mesothelin yields a soluble, polypeptide fragment designated megakaryocyte potentiating factor (MPF) based on its ability to stimulate megakaryocyte colony-forming activity of murine interleukin-3 in murine bone marrow cell cultures. Originally isolated from the HPC-Y5 pancreatic cell line, MPF has been suggested to play a role in the proliferation and differentiation of megakaryoctyes, and the regulation of resultant platelet production. While the biological functions of both Mesothelin and MPF remain speculative, high-levels of expression in cancerous tissues compared to limited distribution in normal tissues strongly suggests their involvement in tumorigenesis. Both have been demonstrated to promote tumor cell proliferation, migration,anchorage-independent growth, and tumor progression; demonstrating their involvement in heterotypic cell adhesion and the metastatic spread of cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mesothelin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q13421
MSLN
Recombinant Human Mesothelin (MSLN), Glu295-Ser597 (Accession Q13421).
20 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
C6
Unconjugated
MSLN
CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; MPF; MSLN; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein
Mouse
Protein A/G
RUO
10232
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.