missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (MSLN/2131), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79858AF750
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal specifically detects Mesothelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Mesothelin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MSLN | |
| Recombinant fragment (around aa 273-407) of human Mesothelin (MSLN) protein (exact sequence is proprietary) (Uniprot: Q13421) | |
| 0.1 mL | |
| Cancer | |
| Human, Mouse, Rat | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MSLN/2131 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction