missing translation for 'onlineSavingsMsg'
Learn More

Mesothelin Antibody (618923) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30738434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738434 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738434 Supplier Novus Biologicals Supplier No. FAB32653MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mesothelin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 618923
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Mesothelin, Glu296-Asp580, Accession # AAH09272
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.