missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesothelin Antibody (420411), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB32652PECY7
This item is not returnable.
View return policy
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry
Specifications
| Mesothelin | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 420411 | |
| Flow Cytometry | |
| CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
| Mouse myeloma cell line NS0-derived recombinant human Mesothelin, Glu296-Gly580, Accession # AAH09272 | |
| 0.1 mL | |
| Cancer | |
| 10232 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction