missing translation for 'onlineSavingsMsg'
Learn More

Mesoridazine, TRC

Product Code. 30005504
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005504 10 mg 10mg
30003632 5 mg 5mg
30006951 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005504 Supplier Toronto Research Chemicals Supplier No. TRCM22576010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5588-33-0
Molecular Formula C21 H26 N2 O S2
Molecular Weight (g/mol) 386.57
Synonym 10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, Phenothiazine, 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)- (7CI,8CI), 1-Methyl-2-[2-[2-(methylsulfinyl)phenothiazin-10-yl]ethyl]piperidine, 10-[2-(1-Methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine, 10-[2-(1-Methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)phenothiazine, Mesoridazine, NSC 186066, Serentil, TPS 23, Thioridazine-2-sulfoxide
IUPAC Name 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfinylphenothiazine
SMILES CN1CCCCC1CCN2c3ccccc3Sc4ccc(cc24)S(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H26N2OS2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(26(2)24)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3
Formula Weight 386.1487
Percent Purity >95%
Chemical Name or Material Mesoridazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.