missing translation for 'onlineSavingsMsg'
Learn More

Mesoridazine.besylate, Lipomed

Product Code. 30458785
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30458785 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30458785

Brand: Lipomed LPMMES1880BS100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Mesoridazine.besylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesoridazine Besilate
CAS 32672-69-8
Analyte or Component Names Mesoridazine Besilate
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 O S2 . C6 H6 O3 S
Synonym Thioridazine Hydrochloride Imp. B (EP), Thioridazine Imp. B (EP), 10 H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, benzenesulfonate (1:1)10 H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, monobenzenesulfonate (9 CI), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10 H-phenothiazine (1:1), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)phenothiazine (1:1) (8 CI), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCCCC1CCN2c3ccccc3Sc4ccc(cc24)S(=O)C.OS(=O)(=O)c5ccccc5
IUPAC Name benzenesulfonic acid;10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfinylphenothiazine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 544.75
Quantity 100 mg
Formula Weight 544.1524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.