missing translation for 'onlineSavingsMsg'
Learn More

Mesoridazine Besilate (10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsufhinyl)-10H-phenothiazine Besilate), Mikromol™

Product Code. 18090180
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090180 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18090180 Supplier Mikromol Supplier No. MM0116.01
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mesoridazine Besilate (10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsufhinyl)-10H-phenothiazine Besilate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesoridazine Besilate
CAS 32672-69-8
Analyte or Component Names Mesoridazine Besilate
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 O S2 . C6 H6 O3 S
Certifications/Compliance ISO 9001
Synonym Thioridazine Hydrochloride Imp. B (EP), Thioridazine Imp. B (EP), 10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, benzenesulfonate (1:1)10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, monobenzenesulfonate (9CI), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine (1:1), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)phenothiazine (1:1) (8CI), Lidanar, Lidanil, Mesoridazine benzenesulfonate, Mesoridazine besylate, NC 123, Imp. B (EP) as Besilate: 10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10H-phenothiazine Besilate (MesoridazineBesilate), Thioridazine Hydrochloride Impurity B as Besilate, Thioridazine Impurity B as Besilate
InChI Formula InChI=1S/C21H26N2OS2.C6H6O3S/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(26(2)24)15-19(21)23;7-10(8,9)6-4-2-1-3-5-6/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1-5H,(H,7,8,9)
SMILES CN1CCCCC1CCN2c3ccccc3Sc4ccc(cc24)S(=O)C.OS(=O)(=O)c5ccccc5
IUPAC Name benzenesulfonic acid;10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfinylphenothiazine
Molecular Weight (g/mol) 544.75
Quantity 50 mg
Formula Weight 544.1524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.