missing translation for 'onlineSavingsMsg'
Learn More

Mesoporphyrin IX dihydrochloride, 97%

Product Code. 11399478
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399478 250 mg 250mg
11389478 10 mg 10mg
11309488 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399478 Supplier Thermo Scientific Alfa Aesar Supplier No. 043320.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68938-72-7 | C34H40Cl2N4O4 | 639.62 g/mol

Mesoporphyrin IX dihydrochloride is used reactant in constructing efficient dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68938-72-7
Molecular Formula C34H40Cl2N4O4
Molecular Weight (g/mol) 639.62
MDL Number MFCD00136049
InChI Key YUIUEEROGVTICN-UHFFFAOYSA-N
Synonym mesoporphyrin ix dihydrochloride, mesoporphyrin ix, dihydrochloride, 7,12-diethyl-3,8,13,17-tetramethyl-21h,23h-porphine-2,18-dipropionic acid dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, hydrochloride 1:2, mesoporphyrin ix dihydrochloride, synthetic, 3-18-2-carboxyethyl-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl propanoic acid dihydrochloride, 3-20-2-carboxyethyl-10,15-diethyl-5,9,14,19-tetramethyl-21,22,23,24-tetraazapentacyclo 16.2.1.1_,?.1?,__.1__,_? tetracosa-1,3 24 ,4,6,8,10,12,14,16 22 ,17,19-undecaen-4-yl propanoic acid dihydrochloride
PubChem CID 111858
SMILES [H+].[H+].[Cl-].[Cl-].CCC1=C(C)C2=CC3=C(CC)C(C)=C(N3)C=C3N=C(C=C4N=C(C=C1N2)C(C)=C4CCC(O)=O)C(CCC(O)=O)=C3C

Specifications

Quantity 250 mg
Solubility Information Soluble in water.
IUPAC Name dihydrogen 3-[20-(2-carboxyethyl)-9,14-diethyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(21),2,4,6(24),7,9,11,13,15,17,19-undecaen-4-yl]propanoic acid dichloride
Formula Weight 639.62
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Mesoporphyrin IX dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.